5-Bromo-3-(trifluoroacetyl)indole

Lieferant: Apollo Scientific

Synonyme: 1-(5-Bromo-1H-indol-3-yl)-2,2,2-trifluoroethanone

PC510142-5G PC510142-1G
APOSPC510142-5GEA 397 EUR
APOSPC510142-5G APOSPC510142-1G
5-Bromo-3-(trifluoroacetyl)indole
5-Bromo-3-(trifluoroacetyl)indole
Formel: C₁₀H₅BrF₃NO
Molecular Weight: 292.052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03721051
CAS-Nummer: 32387-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR