2-Chloro-4'-fluoro-2'-hydroxyacetophenone

Lieferant: Apollo Scientific

PC510143-5G
APOSPC510143-5GEA 1350 EUR
APOSPC510143-5G
2-Chloro-4'-fluoro-2'-hydroxyacetophenone
2-Chloro-4'-fluoro-2'-hydroxyacetophenone
Formel: C₈H₆ClFO₂
Molecular Weight: 188,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22574925
CAS-Nummer: 1410792-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR