Potassium Trifluoro(3-methoxybenzyl)borate

Lieferant: Apollo Scientific

PC510145-1G
APOSPC510145-1GEA 804 EUR
APOSPC510145-1G
Potassium Trifluoro(3-methoxybenzyl)borate
Potassium Trifluoro(3-methoxybenzyl)borate
Formel: C₈H₉BF₃O·K
Molecular Weight: 228,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112716
CAS-Nummer: 1569524-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR