3-Fluoro-N-methoxy-N-methylisonicotinamide

Lieferant: Apollo Scientific

PC510147-5G
APOSPC510147-5GEA 916 EUR
APOSPC510147-5G
3-Fluoro-N-methoxy-N-methylisonicotinamide
3-Fluoro-N-methoxy-N-methylisonicotinamide
Formel: C₈H₉FN₂O₂
Molecular Weight: 184,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21114983
CAS-Nummer: 1187669-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR