6-Bromo-5-fluoro-2-methyl-2H-indazole

Lieferant: Apollo Scientific
Achtung

PC51015-250MG
APOSPC51015-250MGEA 127 EUR
APOSPC51015-250MG
6-Bromo-5-fluoro-2-methyl-2H-indazole
6-Bromo-5-fluoro-2-methyl-2H-indazole
Formel: C₈H₆BrFN₂
Molecular Weight: 229,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089820
CAS-Nummer: 1286734-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR