6-(Allyloxy)-2,3-difluorobenzyl Alcohol
Lieferant: Apollo Scientific
PC510150-5G
APOSPC510150-5GEA
1120
EUR
APOSPC510150-5G
6-(Allyloxy)-2,3-difluorobenzyl Alcohol
6-(Allyloxy)-2,3-difluorobenzyl Alcohol
Formel:
C₁₀H₁₀F₂O₂ Molecular Weight: 200,182 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26937565 CAS-Nummer: 1823942-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...