6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
Lieferant: Apollo Scientific
PC510152-1G
APOSPC510152-1GEA
1120
EUR
APOSPC510152-1G
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
Formel:
C₁₀H₁₀ClFO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD26937567 CAS-Nummer: 1823365-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...