4-Acetamido-3-fluorophenylboronic acid
Lieferant: Apollo Scientific
Synonyme:
4-Acetamido-3-fluorophenylboronic Acid
PC510153-1G
APOSPC510153-1GEA
1120
EUR
APOSPC510153-1G
4-Acetamido-3-fluorophenylboronic acid
4-Acetamido-3-fluorophenylboronic acid
Formel:
C₈H₉BFNO₃ Molecular Weight: 196,971 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26141998 CAS-Nummer: 626251-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...