2-(1,1-Difluoroethyl)benzylamine

Lieferant: Apollo Scientific

PC510154-1G
APOSPC510154-1GEA 1120 EUR
APOSPC510154-1G
2-(1,1-Difluoroethyl)benzylamine
2-(1,1-Difluoroethyl)benzylamine
Formel: C₉H₁₁F₂N
Molecular Weight: 171.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500639
CAS-Nummer: 1638767-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR