2,2-Difluoro-2-phenylethanamine

Lieferant: Apollo Scientific

PC510155-5G
APOSPC510155-5GEA 1220 EUR
APOSPC510155-5G
2,2-Difluoro-2-phenylethanamine
2,2-Difluoro-2-phenylethanamine
Formel: C₈H₉F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD11100799
CAS-Nummer: 55601-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR