Ethyl 2-(2-Bromo-5-fluorophenyl)acetate
Lieferant: Apollo Scientific
PC510156-1G
APOSPC510156-1GEA
601
EUR
APOSPC510156-1G
Ethyl 2-(2-Bromo-5-fluorophenyl)acetate
Ethyl 2-(2-Bromo-5-fluorophenyl)acetate
Formel:
C₁₀H₁₀BrFO₂ Molecular Weight: 261,088 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11554987 CAS-Nummer: 1214910-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...