7-Fluoro-8-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

PC510157-1G
APOSPC510157-1GEA 1120 EUR
APOSPC510157-1G
7-Fluoro-8-hydroxy-2-methylquinoline
7-Fluoro-8-hydroxy-2-methylquinoline
Formel: C₁₀H₈FNO
Molecular Weight: 177,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22688819
CAS-Nummer: 37026-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR