6-Fluoro-8-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

PC510158-1G
APOSPC510158-1GEA 916 EUR
APOSPC510158-1G
6-Fluoro-8-hydroxy-2-methylquinoline
6-Fluoro-8-hydroxy-2-methylquinoline
Formel: C₁₀H₈FNO
Molecular Weight: 177,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19690189
CAS-Nummer: 1070897-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR