5-Bromo-2-chloro-3-fluorophenol

Lieferant: Apollo Scientific

PC510159-1G
APOSPC510159-1GEA 804 EUR
APOSPC510159-1G
5-Bromo-2-chloro-3-fluorophenol
5-Bromo-2-chloro-3-fluorophenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,443 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500640
CAS-Nummer: 186590-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR