3'-(Dimethylamino)-2,2,2-trifluoroacetophenone

Lieferant: Apollo Scientific

Synonyme: 1-(3-(Dimethylamino)phenyl)-2,2,2-trifluoroethanone

PC510160-1G
APOSPC510160-1GEA 916 EUR
APOSPC510160-1G
3'-(Dimethylamino)-2,2,2-trifluoroacetophenone
3'-(Dimethylamino)-2,2,2-trifluoroacetophenone
Formel: C₁₀H₁₀F₃NO
Molecular Weight: 217,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11556187
CAS-Nummer: 33284-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR