4-(2-Iodoethyl)benzotrifluoride
Lieferant: Apollo Scientific
PC510161-1G
APOSPC510161-1GEA
601
EUR
APOSPC510161-1G
4-(2-Iodoethyl)benzotrifluoride
4-(2-Iodoethyl)benzotrifluoride
Formel:
C₉H₈F₃I Molecular Weight: 300,059 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18207440 CAS-Nummer: 178685-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...