2-Amino-6-fluoro-1-methylbenzimidazole

Lieferant: Apollo Scientific

PC510162-1G
APOSPC510162-1GEA 794 EUR
APOSPC510162-1G
2-Amino-6-fluoro-1-methylbenzimidazole
2-Amino-6-fluoro-1-methylbenzimidazole
Formel: C₈H₈FN₃
Molecular Weight: 165,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16841034
CAS-Nummer: 1248916-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR