2-Amino-4-fluoro-1-methylbenzimidazole
Lieferant: Apollo Scientific
PC510163-1G
APOSPC510163-1GEA
916
EUR
APOSPC510163-1G
2-Amino-4-fluoro-1-methylbenzimidazole
2-Amino-4-fluoro-1-methylbenzimidazole
Formel:
C₈H₈FN₃ Molecular Weight: 165,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12149741 CAS-Nummer: 1183473-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...