2-Amino-5-fluoro-1-methylbenzimidazole
Lieferant: Apollo Scientific
PC510164-1G
APOSPC510164-1GEA
794
EUR
APOSPC510164-1G
2-Amino-5-fluoro-1-methylbenzimidazole
2-Amino-5-fluoro-1-methylbenzimidazole
Formel:
C₈H₈FN₃ Molecular Weight: 165,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11215483 CAS-Nummer: 1156928-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...