4-(2,2,2-Trifluoroethoxy)salicylaldehyde

Lieferant: Apollo Scientific

PC510166-5G
APOSPC510166-5GEA 1120 EUR
APOSPC510166-5G
4-(2,2,2-Trifluoroethoxy)salicylaldehyde
4-(2,2,2-Trifluoroethoxy)salicylaldehyde
Formel: C₉H₇F₃O₃
Molecular Weight: 220,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935383
CAS-Nummer: 1823891-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR