2-Bromo-2',6'-difluoro-4'-hydroxyacetophenone

Lieferant: Apollo Scientific

PC510170-1G
APOSPC510170-1GEA 0 EUR
APOSPC510170-1G
2-Bromo-2',6'-difluoro-4'-hydroxyacetophenone
2-Bromo-2',6'-difluoro-4'-hydroxyacetophenone
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18204775
CAS-Nummer: 1379309-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR