Ethyl 2,4-Difluoro-5-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

PC510171-5G
APOSPC510171-5GEA 954 EUR
APOSPC510171-5G
Ethyl 2,4-Difluoro-5-nitrobenzoate
Ethyl 2,4-Difluoro-5-nitrobenzoate
Formel: C₉H₇F₂NO₄
Molecular Weight: 231,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12172523
CAS-Nummer: 179011-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR