4-(1-Amino-2,2,2-trifluoroethyl)phenol

Lieferant: Apollo Scientific

PC510172-1G
APOSPC510172-1GEA 1090 EUR
APOSPC510172-1G
4-(1-Amino-2,2,2-trifluoroethyl)phenol
4-(1-Amino-2,2,2-trifluoroethyl)phenol
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832191
CAS-Nummer: 294175-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR