4-(tert-Butyl)-3,5-difluorophenol

Lieferant: Apollo Scientific

PC510173-1G
APOSPC510173-1GEA 925 EUR
APOSPC510173-1G
4-(tert-Butyl)-3,5-difluorophenol
4-(tert-Butyl)-3,5-difluorophenol
Formel: C₁₀H₁₂F₂O
Molecular Weight: 186,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20485342
CAS-Nummer: 910486-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR