3,5-Difluoro-4-(methylsulphonyl)phenol

Lieferant: Apollo Scientific

PC510174-1G
APOSPC510174-1GEA 1120 EUR
APOSPC510174-1G
3,5-Difluoro-4-(methylsulphonyl)phenol
3,5-Difluoro-4-(methylsulphonyl)phenol
Formel: C₇H₆F₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD27986812
CAS-Nummer: 1447123-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR