3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

PC510176-1G
APOSPC510176-1GEA 79.9 EUR
APOSPC510176-1G
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester
Formel: C₁₃H₁₈BFO₃
Molecular Weight: 252,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18732617
CAS-Nummer: 1082066-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR