3-Amino-2,2-difluoropropanoic acid
Lieferant: Apollo Scientific
PC510177-1G
APOSPC510177-1GEA
1030
EUR
APOSPC510177-1G
3-Amino-2,2-difluoropropanoic acid
3-Amino-2,2-difluoropropanoic acid
Formel:
C₃H₅F₂NO₂ Molecular Weight: 125,074 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864327 CAS-Nummer: 428452-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...