3-(tert-Butoxy)-5-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC510178-1G
APOSPC510178-1GEA 561 EUR
APOSPC510178-1G
3-(tert-Butoxy)-5-(trifluoromethyl)aniline
3-(tert-Butoxy)-5-(trifluoromethyl)aniline
Formel: C₁₁H₁₄F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD24344345
CAS-Nummer: 1369841-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR