3-Cyclopropoxy-5-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC510179-1G
APOSPC510179-1GEA 357 EUR
APOSPC510179-1G
3-Cyclopropoxy-5-(trifluoromethyl)aniline
3-Cyclopropoxy-5-(trifluoromethyl)aniline
Formel: C₁₀H₁₀F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD17008595
CAS-Nummer: 1243471-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR