Methyl 4-(Trifluoromethyl)benzimidate hydrochloride

Lieferant: Apollo Scientific

PC510180-5G
APOSPC510180-5GEA 397 EUR
APOSPC510180-5G
Methyl 4-(Trifluoromethyl)benzimidate hydrochloride
Methyl 4-(Trifluoromethyl)benzimidate hydrochloride
Formel: C₉H₈F₃NO·ClH
Molecular Weight: 239.622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031909
CAS-Nummer: 56108-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR