Methyl 3-(Trifluoromethyl)benzimidate hydrochloride

Lieferant: Apollo Scientific

PC510181-5G
APOSPC510181-5GEA 397 EUR
APOSPC510181-5G
Methyl 3-(Trifluoromethyl)benzimidate hydrochloride
Methyl 3-(Trifluoromethyl)benzimidate hydrochloride
Formel: C₉H₈F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD12031960
CAS-Nummer: 56108-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR