3-(2-Fluoro-3-hydroxyphenyl)pentanoic acid

Lieferant: Apollo Scientific

PC510185-1G
APOSPC510185-1GEA 916 EUR
APOSPC510185-1G
3-(2-Fluoro-3-hydroxyphenyl)pentanoic acid
3-(2-Fluoro-3-hydroxyphenyl)pentanoic acid
Formel: C₁₁H₁₃FO₃
Molecular Weight: 212.218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014570
CAS-Nummer: 1142230-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR