3-Cyclopropyl-3-(2-fluoro-3-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific

PC510186-1G
APOSPC510186-1GEA 916 EUR
APOSPC510186-1G
3-Cyclopropyl-3-(2-fluoro-3-hydroxyphenyl)propanoic acid
3-Cyclopropyl-3-(2-fluoro-3-hydroxyphenyl)propanoic acid
Formel: C₁₂H₁₃FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28014569
CAS-Nummer: 1823386-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR