6-Bromo-5-fluoro-3-iodo-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC51019-100MG
APOSPC51019-100MGEA 21.9 EUR
APOSPC51019-100MG
6-Bromo-5-fluoro-3-iodo-1H-indazole
6-Bromo-5-fluoro-3-iodo-1H-indazole
Formel: C₇H₃BrFIN₂
Molecular Weight: 340.919 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18384871
CAS-Nummer: 1286734-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR