1-(3-Chlorophenyl)-2,2,2-trifluoroethanol

Lieferant: Apollo Scientific

PC510193-1G
APOSPC510193-1GEA 601 EUR
APOSPC510193-1G
1-(3-Chlorophenyl)-2,2,2-trifluoroethanol
1-(3-Chlorophenyl)-2,2,2-trifluoroethanol
Formel: C₈H₆ClF₃O
Molecular Weight: 210,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461901
CAS-Nummer: 81577-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR