1-(4-Chlorophenyl)-2,2,2-trifluoroethanol

Lieferant: Apollo Scientific

PC510194-1G
APOSPC510194-1GEA 561 EUR
APOSPC510194-1G
1-(4-Chlorophenyl)-2,2,2-trifluoroethanol
1-(4-Chlorophenyl)-2,2,2-trifluoroethanol
Formel: C₈H₆ClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00018720
CAS-Nummer: 446-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR