1-(2,6-Difluorophenyl)ethylamine

Lieferant: Apollo Scientific

Synonyme: 1-(2,6-Difluorophenyl)ethanamine

PC510195-5G
APOSPC510195-5GEA 916 EUR
APOSPC510195-5G
1-(2,6-Difluorophenyl)ethylamine
1-(2,6-Difluorophenyl)ethylamine
Formel: C₈H₉F₂N
Molecular Weight: 157,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09041977
CAS-Nummer: 870849-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR