2-(2-Bromo-4-fluoro-6-nitrophenyl)acetic acid

Lieferant: Apollo Scientific

PC510196-1G
APOSPC510196-1GEA 601 EUR
APOSPC510196-1G
2-(2-Bromo-4-fluoro-6-nitrophenyl)acetic acid
2-(2-Bromo-4-fluoro-6-nitrophenyl)acetic acid
Formel: C₈H₅BrFNO₄
Molecular Weight: 278,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101616
CAS-Nummer: 1805151-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR