2,2,2-Trifluoro-N-[3-(hydroxymethyl)phenyl]acetamide
Lieferant: Apollo Scientific
PC510198-5G
APOSPC510198-5GEA
815
EUR
APOSPC510198-5G
2,2,2-Trifluoro-N-[3-(hydroxymethyl)phenyl]acetamide
2,2,2-Trifluoro-N-[3-(hydroxymethyl)phenyl]acetamide
Formel:
C₉H₈F₃NO₂ Molecular Weight: 219,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07330486 CAS-Nummer: 324560-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...