4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]phenylboronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

PC510200-1G
APOSPC510200-1GEA 804 EUR
APOSPC510200-1G
4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]phenylboronic acid pinacol ester
4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]phenylboronic acid pinacol ester
Formel: C₁₈H₂₈BFN₂O₂
Molecular Weight: 334,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28133362
CAS-Nummer: 2096340-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR