4-Fluoro-2-(morpholinomethyl)phenylboronic acid pinacol ester
Lieferant: Apollo Scientific
PC510201-1G
APOSPC510201-1GEA
804
EUR
APOSPC510201-1G
4-Fluoro-2-(morpholinomethyl)phenylboronic acid pinacol ester
4-Fluoro-2-(morpholinomethyl)phenylboronic acid pinacol ester
Formel:
C₁₇H₂₅BFNO₃ Molecular Weight: 321.195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23098826 CAS-Nummer: 2096340-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...