2-Chloro-6,7-difluoroquinoxaline

Lieferant: Apollo Scientific

PC510202-1G
APOSPC510202-1GEA 434 EUR
APOSPC510202-1G
2-Chloro-6,7-difluoroquinoxaline
2-Chloro-6,7-difluoroquinoxaline
Formel: C₈H₃ClF₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11518958
CAS-Nummer: 143007-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR