2-Chloro-7-fluoroquinazoline

Lieferant: Apollo Scientific

PC510203-1G
APOSPC510203-1GEA 191 EUR
APOSPC510203-1G
2-Chloro-7-fluoroquinazoline
2-Chloro-7-fluoroquinazoline
Formel: C₈H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11518968
CAS-Nummer: 956101-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR