Methyl 3-Fluoro-5-hydroxy-4-methoxybenzoate

Lieferant: Apollo Scientific

PC510206-1G
APOSPC510206-1GEA 1840 EUR
APOSPC510206-1G
Methyl 3-Fluoro-5-hydroxy-4-methoxybenzoate
Methyl 3-Fluoro-5-hydroxy-4-methoxybenzoate
Formel: C₉H₉FO₄
Molecular Weight: 200,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16877843
CAS-Nummer: 838856-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR