5-[4-(Trifluoromethoxy)phenyl]nicotinic acid

Lieferant: Apollo Scientific

Synonyme: 5-(4-Trifluoromethoxyphenyl)-nicotinic acid

PC510208-1G
APOSPC510208-1GEA 98.6 EUR
APOSPC510208-1G
5-[4-(Trifluoromethoxy)phenyl]nicotinic acid
5-[4-(Trifluoromethoxy)phenyl]nicotinic acid
Formel: C₁₃H₈F₃NO₃
Molecular Weight: 283,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410279
CAS-Nummer: 898796-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR