2,2-Difluoro-2-(2-fluorophenyl)ethanol

Lieferant: Apollo Scientific

PC510209-1G
APOSPC510209-1GEA 1010 EUR
APOSPC510209-1G
2,2-Difluoro-2-(2-fluorophenyl)ethanol
2,2-Difluoro-2-(2-fluorophenyl)ethanol
Formel: C₈H₇F₃O
Molecular Weight: 176,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28038850
CAS-Nummer: 1380202-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR