3-Bromo-5-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific
Gefahr

PC510210-1G PC510210-250MG PC510210-5G
APOSPC510210-1GEA 601 EUR
APOSPC510210-1G APOSPC510210-250MG APOSPC510210-5G
3-Bromo-5-(trifluoromethyl)benzylamine
3-Bromo-5-(trifluoromethyl)benzylamine
Formel: C₈H₇BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD13185403
CAS-Nummer: 691877-04-0
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR