Methyl 3,3,3-Trifluoro-2-phenylpropionate

Lieferant: Apollo Scientific

PC510211-1G
APOSPC510211-1GEA 997 EUR
APOSPC510211-1G
Methyl 3,3,3-Trifluoro-2-phenylpropionate
Methyl 3,3,3-Trifluoro-2-phenylpropionate
Formel: C₁₀H₉F₃O₂
Molecular Weight: 218,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16617025
CAS-Nummer: 184414-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR