Methyl 6-[2-(Trifluoromethoxy)phenyl]nicotinate
Lieferant: Apollo Scientific
PC510213-1G
APOSPC510213-1GEA
397
EUR
APOSPC510213-1G
Methyl 6-[2-(Trifluoromethoxy)phenyl]nicotinate
Methyl 6-[2-(Trifluoromethoxy)phenyl]nicotinate
Formel:
C₁₄H₁₀F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18408426 CAS-Nummer: 1261782-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...