Methyl 2-Chloro-5-fluoro-6-methylnicotinate

Lieferant: Apollo Scientific

PC510215-250MG
APOSPC510215-250MGEA 397 EUR
APOSPC510215-250MG
Methyl 2-Chloro-5-fluoro-6-methylnicotinate
Methyl 2-Chloro-5-fluoro-6-methylnicotinate
Formel: C₈H₇ClFNO₂
Molecular Weight: 203,598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28252200
CAS-Nummer: 1253383-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR